Assay Details
| Gene Symbol: DOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054988.1 | XP_017910477.1 | 4 - 5 | 1248 | 57 | |
| XM_018054990.1 | XP_017910479.1 | 4 - 5 | 1078 | 57 | ||
| GenBank mRNA | JO577843.1 | - | 4 - 5 | 723 | 57 | |
More Information
Set Membership: |
|

DOK1
Genomic Map