Assay Details
| Gene Symbol: PNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686813.3 | XP_005686870.1 | 6 - 7 | 842 | 72 | |
| XM_018055437.1 | XP_017910926.1 | 5 - 6 | 724 | 72 | ||
| GenBank mRNA | JO577213.1 | - | 1 - 2 | 326 | 72 | |
More Information
Set Membership: |
|

PNO1
Genomic Map