Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064413.1 | XP_017919902.1 | 9 - 10 | 1571 | 66 | |
| XM_018064414.1 | XP_017919903.1 | 10 - 11 | 1845 | 66 | ||
| GenBank mRNA | KA337635.1 | - | 2 - 3 | 1781 | 66 | |
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map