Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693893.2 | XP_005693950.1 | 7 - 8 | 817 | 62 | |
| XM_005693895.2 | XP_005693952.1 | 6 - 7 | 773 | 62 | ||
| XM_018065169.1 | XP_017920658.1 | 8 - 9 | 851 | 62 | ||
| XM_018065170.1 | XP_017920659.1 | 7 - 8 | 817 | 62 | ||
| XM_018065171.1 | XP_017920660.1 | 7 - 8 | 817 | 62 | ||
| GenBank mRNA | JT712148.1 | - | 7 - 8 | 778 | 62 | |
More Information
Set Membership: |
|

TMEM106A
Genomic Map