Assay Details
| Gene Symbol: KHDRBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696178.3 | XP_005696235.2 | 7 - 8 | 949 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KHDRBS2| Gene Symbol: KHDRBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696178.3 | XP_005696235.2 | 7 - 8 | 949 | 85 | |
Set Membership: |
|