Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041245.1 | XP_017896734.1 | 7 - 8 | 1805 | 67 | |
| GenBank mRNA | JO429230.2 | - | 5 - 6 | 1009 | 67 | |
More Information
Set Membership: |
|

DHX32
Genomic Map