Assay Details
| Gene Symbol: LOC102182773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049486.1 | XP_017904975.1 | 1 - 2 | 402 | 72 | |
| XM_018049487.1 | XP_017904976.1 | 1 - 2 | 402 | 72 | ||
More Information
Set Membership: |
|

LOC102182773
Genomic Map