Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049797.1 | XP_017905286.1 | 2 - 3 | 373 | 64 | |
| GenBank mRNA | JO420068.2 | - | 805 | 64 | ||
More Information
Set Membership: |
|

LRPAP1
Genomic Map