Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052323.1 | XP_017907812.1 | 5 - 6 | 2113 | 75 | |
| XM_018052324.1 | XP_017907813.1 | 5 - 6 | 2116 | 75 | ||
| GenBank mRNA | JO380824.2 | - | 316 | 75 | ||
More Information
Set Membership: |
|

RHOBTB2
Genomic Map