Assay Details
| Gene Symbol: PHYHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055941.1 | XP_017911430.1 | 4 - 5 | 588 | 65 | |
| GenBank mRNA | JO410690.2 | - | 7 - 8 | 950 | 65 | |
More Information
Set Membership: |
|

PHYHD1
Genomic Map