Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039462.1 | XP_017894951.1 | 6 - 7 | 4621 | 59 | |
| XM_018039463.1 | XP_017894952.1 | 6 - 7 | 4629 | 59 | ||
| XM_018039464.1 | XP_017894953.1 | 5 - 6 | 4513 | 59 | ||
| XM_018039465.1 | XP_017894954.1 | 4 - 5 | 4459 | 59 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map