Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039509.1 | XP_017894998.1 | 11 - 12 | 4023 | 59 | |
| XM_018039510.1 | XP_017894999.1 | 11 - 12 | 3995 | 59 | ||
| XM_018039511.1 | XP_017895000.1 | 10 - 11 | 3816 | 59 | ||
| XM_018039512.1 | XP_017895001.1 | 10 - 11 | 4343 | 59 | ||
| XM_018039513.1 | XP_017895002.1 | 11 - 12 | 4920 | 59 | ||
| XM_018039514.1 | XP_017895003.1 | 9 - 10 | 4726 | 59 | ||
| XM_018039515.1 | XP_017895004.1 | 9 - 10 | 4684 | 59 | ||
| XM_018039516.1 | XP_017895005.1 | 9 - 10 | 3734 | 59 | ||
| XM_018039517.1 | XP_017895006.1 | 10 - 11 | 4791 | 59 | ||
| GenBank mRNA | JO633179.2 | - | 1 - 2 | 1539 | 59 | |
More Information
Set Membership: |
|

ZNF532
Genomic Map