Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039509.1 | XP_017894998.1 | 10 - 11 | 3873 | 57 | |
| XM_018039510.1 | XP_017894999.1 | 10 - 11 | 3845 | 57 | ||
| XM_018039511.1 | XP_017895000.1 | 9 - 10 | 3666 | 57 | ||
| XM_018039512.1 | XP_017895001.1 | 9 - 10 | 4193 | 57 | ||
| XM_018039513.1 | XP_017895002.1 | 10 - 11 | 4770 | 57 | ||
| XM_018039514.1 | XP_017895003.1 | 8 - 9 | 4576 | 57 | ||
| XM_018039515.1 | XP_017895004.1 | 8 - 9 | 4534 | 57 | ||
| XM_018039516.1 | XP_017895005.1 | 8 - 9 | 3584 | 57 | ||
| GenBank mRNA | JO633179.2 | - | 2 - 3 | 1689 | 57 | |
More Information
Set Membership: |
|

ZNF532
Genomic Map