Assay Details
| Gene Symbol: MPZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046447.1 | XP_017901936.1 | 3 - 4 | 744 | 68 | |
| XM_018046448.1 | XP_017901937.1 | 3 - 4 | 730 | 68 | ||
| XM_018046449.1 | XP_017901938.1 | 3 - 4 | 744 | 68 | ||
| XM_018046450.1 | XP_017901939.1 | 3 - 4 | 745 | 68 | ||
| GenBank mRNA | JT645204.1 | - | 8 - 9 | 1445 | 68 | |
| KA311908.1 | - | 3 - 4 | 2165 | 68 | ||
More Information
Set Membership: |
|

MPZL1
Genomic Map