Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680659.3 | XP_005680716.1 | 7 - 8 | 2764 | 68 | |
| XM_018048284.1 | XP_017903773.1 | 8 - 9 | 2122 | 68 | ||
| GenBank mRNA | JO594058.2 | - | 1 - 2 | 1695 | 68 | |
| KA328837.1 | - | 8 - 9 | 1895 | 68 | ||
More Information
Set Membership: |
|

FOXRED2
Genomic Map