Assay Details
| Gene Symbol: VAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048598.1 | XP_017904087.1 | 3 - 4 | 619 | 68 | |
| GenBank mRNA | JO634971.2 | - | 3 - 4 | 448 | 68 | |
More Information
Set Membership: |
|

VAMP1
Genomic Map