Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053382.1 | XP_017908871.1 | 2 - 3 | 907 | 111 | |
| XM_018053383.1 | XP_017908872.1 | 2 - 3 | 907 | 111 | ||
| GenBank mRNA | JO589218.2 | - | 3 - 4 | 562 | 111 | |
| JO589219.2 | - | 3 - 4 | 562 | 111 | ||
More Information
Set Membership: |
|

FBXO5
Genomic Map