Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691604.2 | XP_005691661.1 | 4 - 5 | 355 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYBB2| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691604.2 | XP_005691661.1 | 4 - 5 | 355 | 79 | |
Set Membership: |
|