Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698500.3 | XP_005698557.2 | 9 - 10 | 3729 | 68 | |
| XM_018041444.1 | XP_017896933.1 | 8 - 9 | 3907 | 68 | ||
More Information
Set Membership: |
|

DCLRE1A
Genomic Map