Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675067.2 | XP_005675124.2 | 4 - 5 | 937 | 70 | |
| XM_013963516.2 | XP_013818970.2 | 3 - 4 | 1250 | 70 | ||
| GenBank mRNA | JO334567.2 | - | 3 - 4 | 1116 | 70 | |
| JO378352.2 | - | 3 - 4 | 639 | 70 | ||
| JO402144.2 | - | 3 - 4 | 1050 | 70 | ||
| JO424470.1 | - | 3 - 4 | 1242 | 70 | ||
| JO430304.2 | - | 3 - 4 | 1182 | 70 | ||
| JO437334.2 | - | 3 - 4 | 1182 | 70 | ||
| JO516165.2 | - | 3 - 4 | 275 | 70 | ||
| JO559790.2 | - | 4 - 5 | 829 | 70 | ||
| JO561930.2 | - | 4 - 5 | 829 | 70 | ||
| JO603366.2 | - | 3 - 4 | 1193 | 70 | ||
| JO631027.2 | - | 3 - 4 | 1143 | 70 | ||
| JR114079.1 | - | 7 - 8 | 1510 | 70 | ||
| JT658573.1 | - | 5 - 6 | 1066 | 70 | ||
| JT705896.1 | - | 4 - 5 | 1500 | 70 | ||
| JT717612.1 | - | 5 - 6 | 1424 | 70 | ||
| JT737783.1 | - | 1 - 2 | 126 | 70 | ||
| KA342463.1 | - | 3 - 4 | 605 | 70 | ||
More Information
Set Membership: |
|

RNF168
Genomic Map