Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053313.1 | XP_017908802.1 | 2 - 3 | 808 | 64 | |
| GenBank mRNA | JO420974.2 | - | 1 - 2 | 624 | 64 | |
More Information
Set Membership: |
|

RAB32
Genomic Map