Assay Details
| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063785.1 | XP_017919274.1 | 5 - 6 | 769 | 64 | |
| XM_018063786.1 | XP_017919275.1 | 5 - 6 | 765 | 64 | ||
| XM_018063787.1 | XP_017919276.1 | 5 - 6 | 763 | 64 | ||
| XM_018063788.1 | XP_017919277.1 | 5 - 6 | 759 | 64 | ||
More Information
Set Membership: |
|

MRPL38
Genomic Map