Assay Details
| Gene Symbol: PELI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054103.1 | XP_017909592.1 | 5 - 6 | 1043 | 56 | |
| XM_018054104.1 | XP_017909593.1 | 5 - 6 | 711 | 56 | ||
| GenBank mRNA | JO433710.2 | - | 5 - 6 | 721 | 56 | |
| JT739541.1 | - | 2 - 3 | 856 | 56 | ||
More Information
Set Membership: |
|

PELI2
Genomic Map