Assay Details
| Gene Symbol: NEURL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041612.1 | XP_017897101.1 | 4 - 5 | 1876 | 53 | |
| XM_018041613.1 | XP_017897102.1 | 4 - 5 | 1393 | 53 | ||
| GenBank mRNA | JO427661.1 | - | 4 - 5 | 1413 | 53 | |
| JO637145.2 | - | 4 - 5 | 1360 | 53 | ||
More Information
Set Membership: |
|

NEURL1
Genomic Map