Assay Details
| Gene Symbol: SHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046646.1 | XP_017902135.1 | 1 - 2 | 729 | 98 | |
| GenBank mRNA | KA327731.1 | - | 1 - 2 | 215 | 98 | |
More Information
Set Membership: |
|

SHH
Genomic Map