Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683829.3 | XP_005683886.1 | 8 - 9 | 906 | 58 | |
| GenBank mRNA | JO437609.2 | - | 8 - 9 | 854 | 58 | |
| KA330114.1 | - | 8 - 9 | 891 | 58 | ||
More Information
Set Membership: |
|

STOML2
Genomic Map