Assay Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686333.2 | XP_005686390.1 | 4 - 5 | 514 | 65 | |
| XM_005686334.3 | XP_005686391.1 | 3 - 4 | 821 | 65 | ||
| XM_005686335.2 | XP_005686392.1 | 4 - 5 | 514 | 65 | ||
| XM_013967486.2 | XP_013822940.1 | 3 - 4 | 451 | 65 | ||
| GenBank mRNA | JT679815.1 | - | 8 - 9 | 1549 | 65 | |
More Information
Set Membership: |
|

WDR54
Genomic Map