Assay Details
| Gene Symbol: TBCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692271.3 | XP_005692328.1 | 5 - 6 | 707 | 68 | |
| XM_013971219.2 | XP_013826673.1 | 5 - 6 | 567 | 68 | ||
| GenBank mRNA | JO563800.2 | - | 5 - 6 | 730 | 68 | |
| KA329435.1 | - | 1 - 2 | 259 | 68 | ||
More Information
Set Membership: |
|

TBCB
Genomic Map