Assay Details
| Gene Symbol: C19H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693228.3 | XP_005693285.1 | 5 - 6 | 710 | 83 | |
| GenBank mRNA | JO594835.2 | - | 5 - 6 | 712 | 83 | |
More Information
Set Membership: |
|

C19H17orf75
Genomic Map