Assay Details
| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698798.3 | XP_005698855.2 | 2 - 3 | 396 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBOAT4| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698798.3 | XP_005698855.2 | 2 - 3 | 396 | 87 | |
Set Membership: |
|