Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044297.1 | XP_017899786.1 | 4 - 5 | 1794 | 63 | |
| GenBank mRNA | JO412911.2 | - | 1 - 2 | 563 | 63 | |
More Information
Set Membership: |
|

CPT2
Genomic Map