Assay Details
| Gene Symbol: MPST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048292.1 | XP_017903781.1 | 2 - 3 | 747 | 108 | |
| XM_018048294.1 | XP_017903783.1 | 2 - 3 | 728 | 108 | ||
| XM_018048295.1 | XP_017903784.1 | 2 - 3 | 695 | 108 | ||
| XM_018048296.1 | XP_017903785.1 | 2 - 3 | 1008 | 108 | ||
| GenBank mRNA | JO433305.2 | - | 2 - 3 | 774 | 108 | |
| JQ923479.1 | - | 1 - 2 | 390 | 108 | ||
More Information
Set Membership: |
|

MPST
Genomic Map