Assay Details
| Gene Symbol: ZNF346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050625.1 | XP_017906114.1 | 6 - 7 | 878 | 60 | |
| XM_018050626.1 | XP_017906115.1 | 6 - 7 | 830 | 60 | ||
| XM_018050627.1 | XP_017906116.1 | 6 - 7 | 756 | 60 | ||
| GenBank mRNA | KA316502.1 | - | 6 - 7 | 706 | 60 | |
More Information
Set Membership: |
|

ZNF346
Genomic Map