Assay Details
Gene Symbol: HACD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051907.1 | XP_017907396.1 | 3 - 4 | 340 | 98 | |
GenBank mRNA | KA308686.1 | - | 3 - 4 | 299 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
