Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061104.1 | XP_017916593.1 | 2 - 3 | 632 | 69 | |
| GenBank mRNA | JO621180.2 | - | 1 - 2 | 1095 | 69 | |
| JO621181.2 | - | 1 - 2 | 1095 | 69 | ||
More Information
Set Membership: |
|

FAM222A
Genomic Map