Assay Details
| Gene Symbol: CALM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062643.1 | XP_017918132.1 | 2 - 3 | 363 | 72 | |
| GenBank mRNA | JO579083.1 | - | 4 - 5 | 2017 | 72 | |
More Information
Set Membership: |
|

CALM3
Genomic Map