Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675872.3 | XP_005675929.2 | 2 - 3 | 404 | 63 | |
| GenBank mRNA | JO414817.2 | - | 2 - 3 | 223 | 63 | |
More Information
Set Membership: |
|

TFPI
Genomic Map