Assay Details
| Gene Symbol: C5H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680023.3 | XP_005680080.2 | 7 - 8 | 441 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H12orf54| Gene Symbol: C5H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680023.3 | XP_005680080.2 | 7 - 8 | 441 | 59 | |
Set Membership: |
|