Assay Details
| Gene Symbol: SEPT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049613.1 | XP_017905102.1 | 5 - 6 | 862 | 62 | |
| XM_018049614.1 | XP_017905103.1 | 5 - 6 | 863 | 62 | ||
| XM_018049615.1 | XP_017905104.1 | 5 - 6 | 863 | 62 | ||
| XM_018049616.1 | XP_017905105.1 | 5 - 6 | 863 | 62 | ||
| XM_018049617.1 | XP_017905106.1 | 5 - 6 | 863 | 62 | ||
| XM_018049618.1 | XP_017905107.1 | 5 - 6 | 862 | 62 | ||
| XM_018049619.1 | XP_017905108.1 | 5 - 6 | 862 | 62 | ||
| XM_018049620.1 | XP_017905109.1 | 5 - 6 | 862 | 62 | ||
More Information
Set Membership: |
|

SEPT11
Genomic Map