Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682696.3 | XP_005682753.2 | 12 - 13 | 1379 | 68 | |
| XM_005682697.3 | XP_005682754.2 | 12 - 13 | 1378 | 68 | ||
| XM_013965354.2 | XP_013820808.2 | 12 - 13 | 1331 | 68 | ||
| XM_013965356.2 | XP_013820810.2 | 12 - 13 | 1328 | 68 | ||
| XM_018050697.1 | XP_017906186.1 | 13 - 14 | 1494 | 68 | ||
| XM_018050698.1 | XP_017906187.1 | 13 - 14 | 1496 | 68 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map