Assay Details
Gene Symbol: PCED1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688214.3 | XP_005688271.2 | 2 - 3 | 514 | 68 | |
XM_005688216.3 | XP_005688273.2 | 2 - 3 | 512 | 68 | ||
XM_005688217.3 | XP_005688274.2 | 2 - 3 | 519 | 68 | ||
GenBank mRNA | JO641238.2 | - | 2 - 3 | 604 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
