Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697763.3 | XP_005697820.1 | 3 - 4 | 2519 | 66 | |
| XM_018040715.1 | XP_017896204.1 | 4 - 5 | 852 | 66 | ||
More Information
Set Membership: |
|

TMEM248
Genomic Map