Assay Details
| Gene Symbol: PHYHIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699048.3 | XP_005699105.1 | 3 - 4 | 833 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHYHIPL| Gene Symbol: PHYHIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699048.3 | XP_005699105.1 | 3 - 4 | 833 | 95 | |
Set Membership: |
|