Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699947.3 | XP_005700004.1 | 2 - 3 | 415 | 59 | |
| GenBank mRNA | JO437967.2 | - | 4 - 5 | 833 | 59 | |
| JO562101.2 | - | 4 - 5 | 769 | 59 | ||
More Information
Set Membership: |
|

AIP
Genomic Map