Assay Details
| Gene Symbol: MBTPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005701028.3 | XP_005701085.2 | 2 - 3 | 367 | 62 | |
| XM_018043875.1 | XP_017899364.1 | 2 - 3 | 369 | 62 | ||
| GenBank mRNA | JT662486.1 | - | 12 - 13 | 2595 | 62 | |
| JT705377.1 | - | 1 - 2 | 77 | 62 | ||
More Information
Set Membership: |
|

MBTPS2
Genomic Map