Assay Details
| Gene Symbol: C5H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963898.2 | XP_013819352.2 | 7 - 8 | 1604 | 99 | |
| GenBank mRNA | KA328402.1 | - | 2 - 3 | 147 | 99 | |
More Information
Set Membership: |
|

C5H12orf56
Genomic Map