Assay Details
| Gene Symbol: C5H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963898.2 | XP_013819352.2 | 8 - 9 | 1700 | 90 | |
| GenBank mRNA | KA328402.1 | - | 3 - 4 | 243 | 90 | |
More Information
Set Membership: |
|

C5H12orf56
Genomic Map