Assay Details
| Gene Symbol: HS1BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055593.1 | XP_017911082.1 | 6 - 7 | 994 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS1BP3| Gene Symbol: HS1BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055593.1 | XP_017911082.1 | 6 - 7 | 994 | 78 | |
Set Membership: |
|