Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065413.1 | XP_017920902.1 | 7 - 8 | 1258 | 66 | |
| XM_018065414.1 | XP_017920903.1 | 7 - 8 | 1259 | 66 | ||
More Information
Set Membership: |
|

CCDC57
Genomic Map