Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065503.1 | XP_017920992.1 | 2 - 3 | 1410 | 104 | |
| GenBank mRNA | JO332847.2 | - | 2 - 3 | 1329 | 104 | |
| JO611932.2 | - | 1 - 2 | 1920 | 104 | ||
More Information
Set Membership: |
|

PTGER4
Genomic Map